Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Tetrafluoroborate 98.0+%, TCI America™
GREENER_CHOICESDP

Catalog No. B2195100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2195100G 100 g
B21955G 5 g
B219525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B2195100G Supplier TCI America Supplier No. B2195100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174501-65-6
Molecular Formula C8H15BF4N2
Molecular Weight (g/mol) 226.03
MDL Number MFCD03095449
InChI Key LSBXQLQATZTAPE-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate
PubChem CID 2734178
IUPAC Name 1-butyl-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 100 g
UN Number 1760
Formula Weight 226.03
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.