Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Trifluoroacetate 97.0+%, TCI America™
SDP

Catalog No. B552625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B552625G 25 g
B55265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B552625G Supplier TCI America Supplier No. B552625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174899-94-6
Molecular Formula C10H15F3N2O2
Molecular Weight (g/mol) 252.237
MDL Number MFCD08458921
InChI Key QPDGLRRWSBZCHP-UHFFFAOYSA-M
PubChem CID 10125280
IUPAC Name 1-butyl-3-methylimidazol-3-ium;2,2,2-trifluoroacetate
SMILES CCCCN1C=C[N+](=C1)C.C(=O)(C(F)(F)F)[O-]

Specifications

Quantity 25 g
Formula Weight 252.24
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.