Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™
SDP

Catalog No. B233725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B233725G 25 g
B23375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B233725G Supplier TCI America Supplier No. B233725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174899-66-2
Molecular Formula C9H15F3N2O3S
Molecular Weight (g/mol) 288.285
MDL Number MFCD03427620
InChI Key FRZPYEHDSAQGAS-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium trifluoromethanesulfonate, 1-butyl-3-methylimidazolium trifluoromethansulfonate, 1-butyl-3-methylimidazolium triflate, c4mim triflate, bmim otf, acmc-1bwln, basionics™ vs 12, dsstox_cid_29088, dsstox_rid_83307, dsstox_gsid_49232
PubChem CID 2734246
IUPAC Name 1-butyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 16°C
Color Yellow
Quantity 25 g
Formula Weight 288.29
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material 1-Butyl-3-methylimidazolium Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.