Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyl-3-methylpyridinium Bromide 98.0+%, TCI America™
SDP

Catalog No. B310425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B310425G 25 g
B31045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B310425G Supplier TCI America Supplier No. B310425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26576-85-2
Molecular Formula C10H16BrN
Molecular Weight (g/mol) 230.15
MDL Number MFCD01632121
InChI Key GNPWBXOERPGDFI-UHFFFAOYSA-M
PubChem CID 12248306
IUPAC Name 1-butyl-3-methylpyridin-1-ium bromide
SMILES [Br-].CCCC[N+]1=CC=CC(C)=C1

Specifications

Quantity 25 g
Formula Weight 230.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-3-methylpyridinium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.