Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylpyridinium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. B219625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B219625G 25 g
B21965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B219625G Supplier TCI America Supplier No. B219625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 186088-50-6
Molecular Formula C9H14F6NP
Molecular Weight (g/mol) 281.18
MDL Number MFCD03093297
InChI Key CTMFVASPBJWPFC-UHFFFAOYSA-N
Synonym 1-butylpyridinium hexafluorophosphate, n-butylpyridinium hexafluorophosphate, 1-butylpyridin-1-ium hexafluorophosphate v, 1-butylpyridin-1-ium hexafluorophosphate, acmc-1c03e, dsstox_cid_27908, dsstox_rid_82661, dsstox_gsid_47932, ksc171g1j
PubChem CID 2734176
IUPAC Name 1-butylpyridin-1-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCC[N+]1=CC=CC=C1

Specifications

Melting Point 75°C
Color White
Quantity 25 g
Formula Weight 281.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butylpyridinium Hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.