Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Carbobenzoxy-3-piperidinecarboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. C28051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28051G 1 g
C28055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28051G Supplier TCI America Supplier No. C28051G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78190-11-1
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD02931282
InChI Key FFLPIVZNYJKKDM-UHFFFAOYSA-N
Synonym 1-benzyloxy carbonyl-3-piperidinecarboxylic acid, 1-benzyloxy carbonyl piperidine-3-carboxylic acid, n-cbz-piperidine-3-carboxylic acid, 1-cbz-3-piperidinecarboxylic acid, 1-n-cbz-piperidine-3-carboxylic acid, n-cbz-3-piperidinecarboxylic acid, n-cbz-dl-nipecotic acid, 1-benzyloxycarbonyl piperidine-3-carboxylic acid, 1-n-cbz-piperidin-3-ylcarboxylic acid, 1,3-piperidinedicarboxylic acid, 1-phenylmethyl ester
PubChem CID 234339
IUPAC Name 1-phenylmethoxycarbonylpiperidine-3-carboxylic acid
SMILES C1CC(CN(C1)C(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 97°C
Color White
Quantity 1 g
Formula Weight 263.29
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Carbobenzoxy-3-piperidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.