Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Carbobenzoxyamino)cyclopropanecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C26801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26801G 1 g
C26805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26801G Supplier TCI America Supplier No. C26801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84677-06-5
Molecular Formula C12H13NO4
Molecular Weight (g/mol) 235.239
MDL Number MFCD00083285
InChI Key KHINKCGJKZSHAJ-UHFFFAOYSA-N
Synonym 1-(Cbz-amino)cyclopropanecarboxylic Acid
PubChem CID 3589012
IUPAC Name 1-(phenylmethoxycarbonylamino)cyclopropane-1-carboxylic acid
SMILES C1CC1(C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 235.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Carbobenzoxyamino)cyclopropanecarboxylic Acid
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.