Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Carbobenzoxypiperazine 95.0+%, TCI America™
SDP

Catalog No. C217825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C217825G 25 g
C21785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C217825G Supplier TCI America Supplier No. C217825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31166-44-6
Molecular Formula C12H17N2O2
Molecular Weight (g/mol) 221.28
MDL Number MFCD00274317
InChI Key CTOUWUYDDUSBQE-UHFFFAOYSA-O
Synonym 1-cbz-piperazine, benzyl 1-piperazinecarboxylate, 1-benzyloxycarbonyl piperazine, 1-carbobenzoxypiperazine, n-cbz-piperazine, 1-z-piperazine, 1-piperazinecarboxylic acid, phenylmethyl ester, piperazine-1-carboxylic acid benzyl ester, phenylmethyl piperazinecarboxylate, cbz-piperazine
PubChem CID 643495
IUPAC Name 4-[(benzyloxy)carbonyl]piperazin-1-ium
SMILES O=C(OCC1=CC=CC=C1)N1CC[NH2+]CC1

Specifications

Boiling Point 161°C
Quantity 25 g
Formula Weight 220.27
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Carbobenzoxypiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.