Learn More
Medchemexpress LLC 1-Cbz-azetidine-3-carboxylic acid | 97628-92-7 | MFCD09027504 | 99.6% | 235.24 g/mol | C12H13NO4 | 10 G
Supplier: Medchemexpress LLC HYW00486810G
1-Cbz-azetidine-3-carboxylic acid is a protected azetidine carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain-based linker in PROTAC synthesis. Supplied as a powder for research use, it offers high purity and defined storage stability.
- Used as a non-cleavable ADC linker and PROTAC linker.
- High purity suitable for synthetic chemistry.
- Available in multiple package sizes including 10 g.
- Powder stable at -20°C for long-term storage.
- Molecular weight 235.24 g/mol and formula C12H13NO4.