Learn More
Medchemexpress LLC 1-Cbz-azetidine-3-carboxylic acid | 97628-92-7 | MFCD09027504 | 99.6% | 235.24 | C12H13NO4 | 25 G
Supplier: Medchemexpress LLC HYW00486825G
1-Cbz-azetidine-3-carboxylic acid is an N-protected azetidine carboxylic acid used as a non-cleavable linker in antibody-drug conjugate (ADC) synthesis and as an alkyl chain linker for PROTAC construction. It is supplied as a white to off-white solid with high purity suitable for medicinal chemistry and bioconjugation applications.
- non-cleavable ADC linker for antibody-drug conjugates
- alkyl chain-based PROTAC linker for targeted protein degradation
- high purity (99.6%) suitable for synthesis and conjugation
- soluble in DMSO at 100 mg/mL; may require ultrasonic
- stable as powder at -20°C for long-term storage, or 4°C for shorter term
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.