Learn More
Sigma Organic Chemistry 1-chlOR-4-fluOR-2-iodob | 250mg | MFCD00042575
Supplier: Sigma Organic Chemistry CDS010723250MG

1-chlOR-4-fluOR-2-iodob , 250mg
About this Item:
MDL #: MFCD00042575
Molecular Weight: 256.44
UNSPSC Code: 12352200
Chemical Formula: C6H3ClFI
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.