Learn More
1-Chloro-4-fluorobenzene, 98%
Description
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4'-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 '-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4′-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 ′-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | -22°C |
| Density | 1.228 |
| Boiling Point | 129°C to 130°C |
| Flash Point | 29°C (84°F) |
| Refractive Index | 1.495 |
| Quantity | 100 g |
| UN Number | UN1993 |
| Beilstein | 1904542 |
| Solubility Information | Difficult to mix. |
| Formula Weight | 130.55 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H332-H315-H319-H335 Flammable liquid and vapour. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 206-521-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.