Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-4-iodobenzene 99.0+%, TCI America™
SDP

Catalog No. C1075250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1075250G 250 g
C107525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1075250G Supplier TCI America Supplier No. C1075250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 637-87-6
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.452
MDL Number MFCD00001053
InChI Key GWQSENYKCGJTRI-UHFFFAOYSA-N
Synonym 4-chloroiodobenzene, p-chloroiodobenzene, 4-iodochlorobenzene, p-iodochlorobenzene, benzene, 1-chloro-4-iodo, 1-chloro-4-iodo-benzene, p-chlorjodbenzol, 4-chloro iodobenzene, 4-chloro-iodobenzene, 4-iodo-chlorobenzene
PubChem CID 12510
IUPAC Name 1-chloro-4-iodobenzene
SMILES C1=CC(=CC=C1Cl)I

Specifications

Melting Point 56°C
Boiling Point 227°C
Quantity 250 g
Formula Weight 238.45
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Chloro-4-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.