Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorobutane CHROMASOLV™, for HPLC, ≥99.8%, Solstice

Catalog No. 6000094 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-000-94 2.5 L
60-000-95 2 L
60-000-93 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-000-94 Supplier Solstice Supplier No. 349582.5L

CHROMASOLV™, for HPLC,≥99.8%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point -123°C
Density 0.886g/mL at 25°C (lit.)
Boiling Point 77°C to 78°C
Refractive Index n20/D 1.402
Linear Formula CH3(CH2)3Cl
Quantity 2.5 L
Beilstein 1730909
Vapor Pressure 80.1mmHg (78.4°C)
Formula Weight 92.57g/mol
Percent Purity ≥99.8%
Grade CHROMASOLV
Chemical Name or Material 1-Chlorobutane, for HPLC
Show More Show Less
Hazard Statement P210-P273-P301 + P310-P331-P403 + P235
Precautionary Statement H225-H304-H336-H412
EINECSNumber 203-696-6
RTECSNumber EJ6300000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.