Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloroethyl chloroformate, 97+%

Catalog No. AC314461000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314461000 100 g Glass bottle
AC314460250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314461000 Supplier Thermo Scientific Chemicals Supplier No. 314461000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 50893-53-3 | C3H4Cl2O2 | 142.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50893-53-3
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000647
InChI Key QOPVNWQGBQYBBP-UHFFFAOYNA-N
Synonym 1-chloroethyl chloroformate, chloroformic acid 1-chloroethyl ester, 1-chloro ethyl chloroformate, 1-chloroethylchloroformate, carbonochloridic acid, 1-chloroethyl ester, ace-cl, alpha-chloroethyl chloroformate, chloro 1-chloroethoxy methanone, .alpha.-chloroethyl chloroformate, pubchem17418
PubChem CID 521305
IUPAC Name 1-chloroethyl carbonochloridate
SMILES CC(Cl)OC(Cl)=O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 1.3200g/mL
Boiling Point 118°C to 119°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2CH(Cl)CH3
Refractive Index 1.4220 to 1.424
Quantity 100 g
Specific Gravity 1.32
Formula Weight 142.97
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material 1-Chloroethyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 256-834-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.