accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50893-53-3

IUPAC Name: 1-chloroethyl carbonochloridate
Synonyms: chloroethyl chloroformate
Molecular Weight (g/mol): 142.96
Molecular Formula: C3H4Cl2O2
InChi Key: QOPVNWQGBQYBBP-UHFFFAOYNA-N
SMILES: CC(Cl)OC(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (2)
  • (2)