Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloromethyl-1H-1,2,4-triazole hydrochloride, 96%

Catalog No. AAH3383003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3383003 1 g
AAH3383006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3383003 Supplier Thermo Scientific Chemicals Supplier No. H3383003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 84387-61-1 | C3H4ClN3 | 117.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84387-61-1
Molecular Formula C3H4ClN3
Molecular Weight (g/mol) 117.54
MDL Number MFCD00964947
InChI Key PXNMFQMREJNQQT-UHFFFAOYSA-N
PubChem CID 3743882
SMILES ClCN1C=NC=N1

Specifications

Melting Point 138°C to 145°C
Quantity 1 g
Sensitivity Hygroscopic
Synonym 1-chloromethyl-1h-1,2,4-triazole, 1-chloromethyl-1,2,4-triazole, 1h-1,2,4-triazole,1-chloromethyl, 1h-1,2,4-triazole, 1-chloromethyl, 1-chloro-methyl-1h-1,2,4-triazole
IUPAC Name 1-(chloromethyl)-1H-1,2,4-triazole
Formula Weight 154
Percent Purity 96%
Chemical Name or Material 1-Chloromethyl-1H-1,2,4-triazole hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.