Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorononane 98.0+%, TCI America™
SDP

Catalog No. C023325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C023325ML 25 mL
C0233500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C023325ML Supplier TCI America Supplier No. C023325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2473-01-0
Molecular Formula C9H19Cl
Molecular Weight (g/mol) 162.701
MDL Number MFCD00001023
InChI Key RKAMCQVGHFRILV-UHFFFAOYSA-N
PubChem CID 17185
IUPAC Name 1-chlorononane
SMILES CCCCCCCCCCl

Specifications

Color Colorless
Boiling Point 204°C
Quantity 25 mL
Synonym nonyl chloride, nonane, 1-chloro, n-nonyl chloride, nonane,1-chloro, tetrapropenyl chloride, 1-propene, tetramer, chlorinated, acmc-209gec, 1-chlorononane, dsstox_cid_29111, dsstox_rid_83330
Formula Weight 162.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorononane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.