Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chlorotetradecane 98.0+%, TCI America™
SDP

Catalog No. C062225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C062225ML 25 mL
C0622500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C062225ML Supplier TCI America Supplier No. C062225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2425-54-9
Molecular Formula C14H29Cl
Molecular Weight (g/mol) 232.836
MDL Number MFCD00000958
InChI Key RNHWYOLIEJIAMV-UHFFFAOYSA-N
Synonym myristyl chloride, tetradecane, 1-chloro, tetradecyl chloride, n-tetradecyl chloride, tetradecane, chloro, chlorotetradecane, 1-chloranyltetradecane, tetradecylchloride, 1-chloro-tetradecane, acmc-1cmeh
PubChem CID 17043
IUPAC Name 1-chlorotetradecane
SMILES CCCCCCCCCCCCCCCl

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
Formula Weight 232.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Chlorotetradecane
EINECSNumber (2)-0066&(2)-0068
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.