Learn More
Sigma Organic Chemistry 1-cyclohexen-1-Y-boroni | 100mg | MFCD02179497
Supplier: Sigma Organic Chemistry CDS004318100MG

1-cyclohexen-1-Y-boroni , 100mg
About this Item:
MDL #: MFCD02179497
Molecular Weight: 125.96
UNSPSC Code: 12352200
Chemical Formula: C6H11BO2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.