Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexene-1,2-dicarboxylic Anhydride 98.0+%, TCI America™
SDP

Catalog No. C170125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C170125G 25 g
C17015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C170125G Supplier TCI America Supplier No. C170125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2426-02-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005917
InChI Key HMMBJOWWRLZEMI-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione
PubChem CID 12475
IUPAC Name 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=C1CCCC2

Specifications

Melting Point 72°C
Color White
Quantity 25 g
UN Number 2698
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyclohexene-1,2-dicarboxylic Anhydride
EINECSNumber (3)-3435
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.