Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexene-1-carboxylic acid, 96%

Catalog No. ac291750025
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC291750025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC291750025 Supplier Thermo Scientific Chemicals Supplier No. 291750025
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 636-82-8 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-82-8
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00001545
InChI Key NMEZJSDUZQOPFE-UHFFFAOYSA-N
Synonym 1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid
PubChem CID 69470
ChEBI CHEBI:29565
IUPAC Name cyclohexene-1-carboxylic acid
SMILES C1CCC(=CC1)C(=O)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 35.0°C to 39.0°C
Color White to Yellow
Boiling Point 137.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5
Quantity 2.5 g
Formula Weight 126.16
Percent Purity 96%
Physical Form Crystalline Mass or Liquid
Chemical Name or Material 1-Cyclohexene-1-carboxylic acid, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.