Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexylethanol, 98%

Catalog No. AAB2036306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2036306 5 g
AAB2036314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2036306 Supplier Thermo Scientific Chemicals Supplier No. B2036306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1193-81-3 | C8H16O | 128.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-81-3
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001475
InChI Key JMSUNAQVHOHLMX-UHFFFAOYSA-N
Synonym methylcyclohexylcarbinol, 1-cyclohexyl-1-ethanol, cyclohexylmethylcarbinol, 1-cyclohexylethan-1-ol, ethanol, 1-cyclohexyl, methanol, cyclohexylmethyl, cyclohexanemethanol, .alpha.-methyl, +--1-cyclohexylethanol, 1-cyclohexyl-ethanol, acmc-20aovg
PubChem CID 137829
IUPAC Name 1-cyclohexylethanol
SMILES CC(C1CCCCC1)O

Specifications

Density 0.92
Boiling Point 188°C to 190°C
Flash Point 73°C (163°F)
Refractive Index 1.465
Quantity 5 g
Formula Weight 128.22
Percent Purity 98%
Chemical Name or Material 1-Cyclohexylethanol
Hazard Category H227-H312-H315-H319
Hazard Statement GHS H Statement
H301-H311-H227
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-780-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.