Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexylpiperazine 98.0+%, TCI America™
SDP

Catalog No. C178325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C178325G 25 g
C17835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C178325G Supplier TCI America Supplier No. C178325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17766-28-8
Molecular Formula C10H20N2
Molecular Weight (g/mol) 168.284
MDL Number MFCD00044809
InChI Key XPDSXKIDJNKIQY-UHFFFAOYSA-N
Synonym n-cyclohexylpiperazine, 1-cyclohexyl-piperazine, cyclohexylpiperazine, piperazine, 1-cyclohexyl, 1-cyclohexylpiperazin, n-cyclohexyl piperazine, 1-cyclohexyl piperazine, piperazine,1-cyclohexyl, 4-1-cyclohexyl piperazine, acmc-1c47v
PubChem CID 87298
IUPAC Name 1-cyclohexylpiperazine
SMILES C1CCC(CC1)N2CCNCC2

Specifications

Melting Point 36°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Cyclohexylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.