Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopentenecarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C17021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17021G 1 g
C17025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17021G Supplier TCI America Supplier No. C17021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1560-11-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001396
InChI Key PYRZPBDTPRQYKG-UHFFFAOYSA-N
Synonym 1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
PubChem CID 95964
IUPAC Name cyclopentene-1-carboxylic acid
SMILES C1CC=C(C1)C(=O)O

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 112.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyclopentenecarboxylic Acid
RTECSNumber GY5976100
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.