Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C27795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27795G 5 g
C27791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27795G Supplier TCI America Supplier No. C27795G

Chemical Identifiers

CAS 93107-30-3
Molecular Formula C13H9F2NO3
Molecular Weight (g/mol) 265.216
MDL Number MFCD01646375
InChI Key KNEXGVPHPGXAGF-UHFFFAOYSA-N
PubChem CID 5210422
IUPAC Name 1-cyclopropyl-6,7-difluoro-4-oxoquinoline-3-carboxylic acid
SMILES C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)F)F)C(=O)O

Specifications

Melting Point 289°C
Color White
Quantity 5 g
Formula Weight 265.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.