Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decanesulfonic Acid Sodium Salt, HPLC Ion Pair Agent, Fisher Chemical™

Catalog No. D003025 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D003025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D003025

missing translation for 'validMainframeMsg'

Improve HPLC separation of basic analytes by ion pairing.

  • High purity
  • Application tested
  • Dissolves easily
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13419-61-9
Molecular Formula C10H21NaO3S
Molecular Weight (g/mol) 244.32
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
IUPAC Name sodium decane-1-sulfonate
SMILES [Na+].CCCCCCCCCCS([O-])(=O)=O

Specifications

Absorbance <0.05 AU at 254 nm
Assay Percent Range 98% min
Quantity 25 g
Grade HPLC
Physical Form Powder
Recommended Storage Ambient
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.