Learn More
Sigma Organic Chemistry 1-Decanethiol | 1G | 143-10-2 | MFCD00004884 | 0.99
Supplier: Sigma Organic Chemistry 7052331G

1-Decanethiol, 1G
About This Item:
Synonym(s): DT; Decyl mercaptan; Mercaptan C10
Linear Formula: CH3(CH2)9SH
Molecular Weight: 174.35
CAS Number: 143-10-2
MDL Number: MFCD00004884
UNSPSC Code: 12352300
Purity: 0.99
Boiling Point: 114 C/13 mmHg (lit.)
Melting Point: -26 C (lit.)
Density: 0.824 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.457 (lit.); n20/D 1.458 (lit.)
Storage: 2-8C
EINECS Number: 205-584-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.