Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decyl-3-methylimidazolium Bromide 98.0+%, TCI America™
SDP

Catalog No. D535025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D535025G 25 g
D53505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D535025G Supplier TCI America Supplier No. D535025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 188589-32-4
Molecular Formula C14H27BrN2
Molecular Weight (g/mol) 303.29
MDL Number MFCD09038832
InChI Key HOISBTKPPVRFDS-UHFFFAOYSA-M
Synonym 1-Decyl-3-methyl-1H-imidazol-3-ium Bromide
PubChem CID 22078297
IUPAC Name 1-decyl-3-methyl-1H-imidazol-3-ium bromide
SMILES [Br-].CCCCCCCCCCN1C=C[N+](C)=C1

Specifications

Quantity 25 g
Formula Weight 303.29
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Decyl-3-methylimidazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.