Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecanethiol, 98%

Catalog No. p-7020696
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1274122 100 g
AAA1274136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1274122 Supplier Thermo Scientific Chemicals Supplier No. A1274122
Only null left
Add to Cart

CAS: 112-55-0 | C12H26S | 202.40 g/mol

1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.

Solubility
Immiscible with water.

Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-55-0
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00004885
InChI Key WNAHIZMDSQCWRP-UHFFFAOYSA-N
Synonym 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
PubChem CID 8195
IUPAC Name dodecane-1-thiol
SMILES CCCCCCCCCCCCS

Specifications

Melting Point °C to 7°C
Density 0.845
Boiling Point 276°C to 278°C
Flash Point 133°C (271°F)
Assay Percent Range 98%
Refractive Index 1.4601
Linear Formula CH3(CH2)11SH
Quantity 100 g
Beilstein 969337
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 202.4
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Dodecanethiol
Show More Show Less
Hazard Category H314-H317-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Precautionary Statement P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 203-984-1
RTECSNumber JR3155000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.