Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecanol, ACS Reagent, ≥98%, Solstice

Catalog No. 60046899 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-899 500 g Poly Bottle
60-046-898 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-899 Supplier Solstice Supplier No. 443816500G

ACS Reagent,≥98.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-53-8
Molecular Formula C12H26O
Molecular Weight (g/mol) 186.34
MDL Number MFCD00004753
InChI Key LQZZUXJYWNFBMV-UHFFFAOYSA-N
Synonym 1-dodecanol, dodecyl alcohol, lauryl alcohol, dodecanol, n-dodecyl alcohol, lauric alcohol, laurinic alcohol, undecyl carbinol, dodecylalcohol, 1-dodecyl alcohol
PubChem CID 8193
ChEBI CHEBI:28878
IUPAC Name dodecan-1-ol
SMILES CCCCCCCCCCCCO

Specifications

Melting Point 22°C to 26°C
Density 0.833g/mL at 25°C
Boiling Point 260°C to 262°C
Flash Point 121°C (249.8°F)
Infrared Spectrum Conforms to structure
Packaging Poly Bottle
Refractive Index n20/D 1.442
Linear Formula CH3(CH2)11OH
Quantity 500 g
UN Number UN3077
Beilstein 1738860
Vapor Density 7.4 (vs air)
Formula Weight 186.33g/mol
Percent Purity ≥98%
Chemical Name or Material 1-Dodecanol
Show More Show Less
Hazard Statement P305 + P351 + P338
DOTInformation 9/PGIII
RTECSNumber 203-982-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.