Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecylamine, 97%

Catalog No. AAA1551536
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1551536 500 g
AAA1551522 100 g
AAA15515A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1551536 Supplier Thermo Scientific Chemicals Supplier No. A1551536
Add to Cart

CAS: 124-22-1 | C12H27N | 185.36 g/mol

Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

Solubility
Soluble in water (78 mg/L 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-22-1
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.36
MDL Number MFCD00008154
InChI Key JRBPAEWTRLWTQC-UHFFFAOYSA-N
Synonym dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
PubChem CID 13583
IUPAC Name dodecan-1-amine
SMILES CCCCCCCCCCCCN

Specifications

Melting Point 27°C to 31°C
Density 0.802
Boiling Point 64°C to 66°C (0.05 mmHg)
Flash Point 115°C (239°F)
Assay Percent Range ≥98%
Quantity 500 g
UN Number UN3259
Beilstein 1633576
Solubility Information Soluble in water (78mg/L 25°C).
Formula Weight 185.36
Percent Purity ≥98%
Chemical Name or Material 1-Dodecylamine
Show More Show Less
Hazard Category H304-H314-H335-H373
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 204-690-6
RTECSNumber JR6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.