Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecyne 95.0+%, TCI America™
SDP

Catalog No. D099725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D099725ML 25 mL
D09975ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D099725ML Supplier TCI America Supplier No. D099725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 765-03-7
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00008960
InChI Key ZVDBUOGYYYNMQI-UHFFFAOYSA-N
Synonym 1-dodecyne, dodecyne, dodec-1-in, 1dodecyne, dodec-1-yn, 1-dodecyne 5g
PubChem CID 69821
IUPAC Name dodec-1-yne
SMILES CCCCCCCCCCC#C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 mL
Formula Weight 166.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Dodecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.