Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one, Thermo Scientific™

Catalog No. AC186310010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC186310010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC186310010 Supplier Thermo Scientific Chemicals Supplier No. 186310010
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10045-45-1
Molecular Formula C9H10N2O
Molecular Weight (g/mol) 162.19
MDL Number MFCD00005715
InChI Key CXUCKELNYMZTRT-UHFFFAOYSA-N
IUPAC Name 1-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
SMILES CCN1C(=O)NC2=CC=CC=C12

Specifications

Alpha Vector ETHYLDIHYDROBENZIMIDAZOLONE
Melting Point 120°C to 126°C
Color White to beige
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 1 g
Physical Form Powder
Chemical Name or Material 1-Ethyl-1,3-dihydro-2H-benzimidazol-2-one
Hazard Category Exclamation mark
EINECSNumber 233-148-1
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.