accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOHN

CAS RN 10045-45-1

IUPAC Name: 1-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-ethyl-2-benzimidazolinone
Molecular Weight (g/mol): 162.19
Molecular Formula: C9H10N2O
InChi Key: CXUCKELNYMZTRT-UHFFFAOYSA-N
SMILES: CCN1C(=O)NC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results