Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Ethyl-1-methylpyrrolidinium bromide, 98%

Click to view available options
Quantity:
25g
100g
69227-51-6
C7H16BrN
194.116
MFCD03095384
KHJQQUGSPDBDRM-UHFFFAOYSA-M
1-ethyl-1-methylpyrrolidinium bromide,1-ethyl-1-methylpyrrolidin-1-ium bromide,1-ethyl-1-methylpyrrolidiniumbromide,pyrrolidinium, 1-ethyl-1-methyl-, bromide,1-methyl-1-ethylpyrrolidinium bromide,pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1,dsstox_cid_29306,dsstox_rid_83422,dsstox_gsid_49347,ksc495s4d
112264
1-ethyl-1-methylpyrrolidin-1-ium;bromide
CC[N+]1(CCCC1)C.[Br-]
69227-51-6
C7H16BrN
194.116
MFCD03095384
KHJQQUGSPDBDRM-UHFFFAOYSA-M
1-ethyl-1-methylpyrrolidinium bromide,1-ethyl-1-methylpyrrolidin-1-ium bromide,1-ethyl-1-methylpyrrolidiniumbromide,pyrrolidinium, 1-ethyl-1-methyl-, bromide,1-methyl-1-ethylpyrrolidinium bromide,pyrrolidinium, 1-ethyl-1-methyl-, bromide 1:1,dsstox_cid_29306,dsstox_rid_83422,dsstox_gsid_49347,ksc495s4d
112264
1-ethyl-1-methylpyrrolidin-1-ium;bromide
CC[N+]1(CCCC1)C.[Br-]
Applications
1-Ethyl-1-methylpyrrolidinium bromide s an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Quantity 25g
Beilstein 14507266
Formula Weight 194.12
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1-Ethyl-1-methylpyrrolidinium bromide

GHS H Statement
H341
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA