Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2-phenylindole 98.0+%, TCI America™
SDP

Catalog No. E033125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E033125G 25 g
E0331500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E033125G Supplier TCI America Supplier No. E033125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13228-39-2
Molecular Formula C16H15N
Molecular Weight (g/mol) 221.303
MDL Number MFCD00031481
InChI Key RAKSXVONTIQCGY-UHFFFAOYSA-N
Synonym 1-ethyl-2-phenyl-1h-indole, n-ethyl-2-phenylindole, pubchem7337, 2-phenyl-1-ethylindole, acmc-1chnt, n-ethyl 2-phenyl indole, 1h-indole,1-ethyl-2-phenyl, 1h-indole, 1-ethyl-2-phenyl
PubChem CID 83248
IUPAC Name 1-ethyl-2-phenylindole
SMILES CCN1C2=CC=CC=C2C=C1C3=CC=CC=C3

Specifications

Melting Point 86°C
Color White-Yellow
Boiling Point 209°C
Quantity 25 g
Formula Weight 221.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-2-phenylindole
EINECSNumber (5)-0138
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.