Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-fluorobenzene, 97%, Thermo Scientific™

Catalog No. AAL0594006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0594006 5 g
AAL0594014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0594006 Supplier Thermo Scientific Chemicals Supplier No. L0594006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Ethyl-4-fluorobenzene is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-47-2
Molecular Formula C8H9F
Molecular Weight (g/mol) 124.16
MDL Number MFCD00039218
InChI Key BLDNWXVISIXWKZ-UHFFFAOYSA-N
Synonym p-ethylfluorobenzene, 4-ethylfluorobenzene, ethyl-4-fluorobenzene, p-fluoroethylbenzene, benzene, 1-ethyl-4-fluoro, 4-fluoroethylbenzene, p-ethyl fluorobenzene, 4-fluoro-ethylbenzene, 1,4-ethylfluorobenzene, pubchem22752
PubChem CID 136303
IUPAC Name 1-ethyl-4-fluorobenzene
SMILES CCC1=CC=C(F)C=C1

Specifications

Boiling Point 139°C
Flash Point 30°C (86°F)
Refractive Index 1.471
Quantity 5 g
UN Number UN1993
Beilstein 1856987
Solubility Information Not miscible in water.
Formula Weight 124.16
Percent Purity 97%
Chemical Name or Material 1-Ethyl-4-fluorobenzene
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.