Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-(methoxycarbonyl)pyridinium Iodide 97.0+%, TCI America™
SDP

Catalog No. E03621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03621G 1 g
E03625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03621G Supplier TCI America Supplier No. E03621G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1199-65-1
Molecular Formula C9H12INO2
Molecular Weight (g/mol) 293.104
MDL Number MFCD00011996
InChI Key NGEAJXXGUZQCPN-UHFFFAOYSA-M
Synonym 4-Carbomethoxy-1-ethylpyridinium Iodide
PubChem CID 70984
IUPAC Name methyl 1-ethylpyridin-1-ium-4-carboxylate;iodide
SMILES CC[N+]1=CC=C(C=C1)C(=O)OC.[I-]

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 293.10
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-4-(methoxycarbonyl)pyridinium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.