Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyladamantane 98.0+%, TCI America™
SDP

Catalog No. E04175ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04175ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04175ML Supplier TCI America Supplier No. E04175ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 770-69-4
Molecular Formula C12H20
Molecular Weight (g/mol) 164.292
MDL Number MFCD00142626
InChI Key LXTHCCWEYOKFSR-UHFFFAOYSA-N
PubChem CID 522637
IUPAC Name 1-ethyladamantane
SMILES CCC12CC3CC(C1)CC(C3)C2

Specifications

Color Colorless
Boiling Point 229°C
Quantity 5 mL
Formula Weight 164.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethyladamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.