Learn More
Sigma Organic Chemistry 1-Ethylpropyl methylami | 1G | 130985-81-8 | MFCD08689639
Supplier: Sigma Organic Chemistry CBR012651G

1-Ethylpropyl methylami , 1G
About this Item:
CAS #: 130985-81-8
MDL #: MFCD08689639
Molecular Weight: 137.65
UNSPSC Code: 12352200
Chemical Formula: C6H16ClN
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.