Learn More
Sigma Organic Chemistry 1-Ethynylpyrene | 100mg | 34993-56-1 | MFCD02093933
Supplier: Sigma Organic Chemistry CDS000782100MG

1-Ethynylpyrene , 100mg
About this Item:
CAS #: 34993-56-1
MDL #: MFCD02093933
Molecular Weight: 226.27
UNSPSC Code: 12352200
Chemical Formula: C18H10
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.