Learn More
1-Fluoro-2-nitrobenzene, 99%
Description
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Fluoro-2-nitrobenzene is an intermediate in the synthetic preparations of various pharmaceutical goods.
Solubility
Immiscible in water.
Notes
Avoid excessive heat and light.
Specifications
Specifications
| Melting Point | -8°C |
| Density | 1.34 |
| Boiling Point | 215°C |
| Flash Point | 94°C (201°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.5317 |
| Quantity | 1000 g |
| UN Number | UN2810 |
| Beilstein | 607262 |
| Solubility Information | Immiscible in water. |
| Show More |
Safety and Handling
| Hazard Category | H301-H310-H315-H319-H373 |
| Hazard Statement | GHS H Statement H301-H311-H331-H373-H315-H319 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P262-P264b-P270-P280-P301+P310-P302+P350-P305+P351+P338-P310-P314-P330-P332+P313-P361-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: II |
| EINECSNumber | 216-088-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.