Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Fluoroadamantane 98.0+%, TCI America™
SDP

Catalog No. F05111G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05111G 1 g
F0511250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05111G Supplier TCI America Supplier No. F05111G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 768-92-3
Molecular Formula C10H15F
Molecular Weight (g/mol) 154.228
InChI Key CPWSNJSGSXXVLD-UHFFFAOYSA-N
Synonym 1-Adamantyl Fluoride
PubChem CID 136608
IUPAC Name 1-fluoroadamantane
SMILES C1C2CC3CC1CC(C2)(C3)F

Specifications

Melting Point 257°C
Color White
Quantity 1 g
Formula Weight 154.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Fluoroadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.