Learn More
Sterilis Solutions 1 Gallon Sharps Container

Description
- Used to comply with FDA and OSHA standards for sharps and biohazard waste collection.
- General-purpose sharps container with clear locking lid and marked with biohazard label.
- Puncture-resistant with wide rectangular opening for vertical and horizontal loading of needles.
- Made from transparent red puncture-resistant polypropylene that can be autoclaved or incinerated.
- Nests to conserve storage space
Specifications
Specifications
| Certifications/Compliance | ISO23907-1:2019 and ISO23908 |
| Closure Type | Slide Entry |
| Closure Color | White/Translucent |
| Capacity (English) | 1 gal. |
| Color | Red |
| Dimensions (L x W x H) | 12.2 x 12.2 x 3.5 in. |
| Disposable | Single-use |
| Latex Free | Yes |
| Quantity | 20/Cs. |
| Sterility | Non-sterile |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.