Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Heptyn-3-ol 97.0+%, TCI America™
SDP

Catalog No. H04551ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04551ML 1 mL
H04555ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04551ML Supplier TCI America Supplier No. H04551ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7383-19-9
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00041597
InChI Key SHSFXAVQBIEYMK-UHFFFAOYSA-N
Synonym 1-heptyn-3-ol, n-butyl ethynyl carbinol, acmc-1biv8, 1-heptyn-3-ol, +/-
PubChem CID 93050
IUPAC Name hept-1-yn-3-ol
SMILES CCCCC(C#C)O

Specifications

Color Yellow
Boiling Point 73°C
Quantity 1 mL
Formula Weight 112.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Heptyn-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.