Learn More
Sigma Organic Chemistry 1-Hexadecanethiol | 500MG | 2917-26-2 | MFCD00011677 | 0.99
Supplier: Sigma Organic Chemistry 674516500MG

1-Hexadecanethiol, 500MG
About This Item:
Synonym(s): Cetyl mercaptan; Hexadecyl mercaptan; Mercaptan C16
Linear Formula: CH3(CH2)15SH
Molecular Weight: 258.51
CAS Number: 2917-26-2
MDL Number: MFCD00011677
UNSPSC Code: 12352103
Purity: 0.99
Boiling Point: 184-191 C/7 mmHg (lit.)
Melting Point: 18-20 C (lit.); 20-24 C
Density: 0.84 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.462 (lit.)
Storage: 2-8C
EINECS Number: 220-846-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.