Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%

Catalog No. AC226991000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC226991000 100 g
AC226995000 500 g
AC226990025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226991000 Supplier Thermo Scientific Chemicals Supplier No. 226991000
Add to Cart

CAS: 6004-24-6 | C21H40ClNO | 358.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6004-24-6
Molecular Formula C21H40ClNO
Molecular Weight (g/mol) 358.01
MDL Number MFCD00149977
InChI Key NFCRBQADEGXVDL-UHFFFAOYSA-M
Synonym cetylpyridinium chloride monohydrate, 1-hexadecylpyridin-1-ium chloride hydrate, 1-hexadecylpyridinium chloride monohydrate, hexadecylpyridinium chloride monohydrate, unii-d9om4sk49p, pyridinium, 1-hexadecyl-, chloride, monohydrate, d9om4sk49p, cetylpyridinii chloridum, sprol tn
PubChem CID 22324
ChEBI CHEBI:3566
IUPAC Name 1-hexadecylpyridin-1-ium;chloride;hydrate
SMILES O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 80.0°C to 83.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96.0-101.0%
Packaging Plastic bottle
Quantity 100 g
Beilstein 20, V,5, 233
Merck Index 15, 2031
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and chloroform, slightly soluble in benzene and ether
Formula Weight 358.01
Percent Purity 96 to 101%
Physical Form Powder
Chemical Name or Material 1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes serious eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occur
RTECSNumber UU5075000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.