Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexanesulfonic Acid Sodium Salt (HPLC), Fisher Chemical™

Catalog No. O3072100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O3072100 100 g
O3072-25 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O3072100
  • $1,149.00
    $858.65 / Each

    Save $290.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $5,530.00
    $4,173.65 / Case of 6 EA

    Save $1,356.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Improve HPLC separation of basic analytes by ion pairing.

  • High purity
  • Application tested
  • Dissolves easily
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
PubChem CID 23677630
IUPAC Name sodium hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Alpha Vector 1-Hexanesulfonic Acid Sodium Salt
Melting Point 300°C
Color White
Assay Percent Range ≥98 %
Linear Formula CH3(CH2)5SO3Na
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 188.22
Percent Purity ≥98%
Grade HPLC
Physical Form Powder or Solid
Chemical Name or Material 1-Hexanesulfonic Acid Sodium Salt
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.