Learn More
(1-Hexyl)triphenylphosphonium bromide, 98%
Description
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
Specifications
Specifications
| Melting Point | 199°C to 202°C |
| Quantity | 100 g |
| Beilstein | 3580731 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water and methanol. |
| IUPAC Name | hexyl(triphenyl)phosphanium;bromide |
| Formula Weight | 427.37 |
| Percent Purity | 98% |
| Chemical Name or Material | (1-Hexyl)triphenylphosphonium bromide |
Safety and Handling
| Hazard Category | H302-H315-H319-H335-H500 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 225-301-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.